missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisomycin, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Anisomycin |
| CAS | 22862-76-6 |
| Molecular Formula | C14H19NO4 |
| Quantity | 50 mg |
| Synonym | (2R,3S,4S)-2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-Acetate, (-)-Anisomycin(2R,3S,4S)-2-(p-Methoxybenzyl)-3,4-pyrrolidinediol 3-Acetate, (2R,3S,4S)-2-(p-Methoxyphenylmethyl)-3-acetoxy-4-hydroxypyrrolidine, (-)-Anisomycin, Flagecidin, NSC 76712, Wuningmeisu C |
| InChI Formula | InChI=1S/C14H19NO4/c1-9(16)19-14-12(15-8-13(14)17)7-10-3-5-11(18-2)6-4-10/h3-6,12-15,17H,7-8H2,1-2H3/t12-,13+,14+/m1/s1 |
| SMILES | COc1ccc(C[C@H]2NC[C@H](O)[C@H]2OC(=O)C)cc1 |
| IUPAC Name | [(2R,3S,4S)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl] acetate |
| Molecular Weight (g/mol) | 265.3 |
| Formula Weight | 265.1314 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction