missing translation for 'onlineSavingsMsg'
Learn More

Anisole, TRC

Product Code. 30332464
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332464 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332464 Supplier Toronto Research Chemicals Supplier No. TRCA673900500G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anisole
CAS 100-66-3
Purity Grade Notes HPLC
Molecular Formula C7 H8 O
Quantity 500 g
Synonym Anisole (8CI), Methoxybenzene, Anisol, Hichemix MB, Methyl phenyl ether, NSC 7920, Phenoxymethane, Phenyl methyl ether
InChI Formula InChI=1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3
SMILES COc1ccccc1
IUPAC Name anisole
Molecular Weight (g/mol) 108.14
Formula Weight 108.0575
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.