missing translation for 'onlineSavingsMsg'
Learn More

Anisole, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 11934721
27.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
1 L
100-66-3
C7H8O
108.14
RDOXTESZEPMUJZ-UHFFFAOYSA-N
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
7519
CHEBI:16579
anisole
COC1=CC=CC=C1
100-66-3
C7H8O
108.14
RDOXTESZEPMUJZ-UHFFFAOYSA-N
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
7519
CHEBI:16579
anisole
COC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -37.0°C
Density 0.9900g/mL
Boiling Point 154.0°C
Flash Point 43°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula C6H5OCH3
Refractive Index 1.5160 to 1.5190
Quantity 100 mL
Merck Index 15,662
Specific Gravity 0.99
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
Formula Weight 108.14
Percent Purity 99%
Grade Extra Dry
Chemical Name or Material Anisole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'