missing translation for 'onlineSavingsMsg'
Learn More

Anisoin, 97%

Product Code. p-7025855
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11487144

Brand: Thermo Scientific Alfa Aesar B21559.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-52-8 | C16H16O4 | 272.30 g/mol

Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

Solubility
Soluble in water, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-52-8
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD00008411
InChI Key LRRQSCPPOIUNGX-UHFFFAOYNA-N
Synonym anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
PubChem CID 95415
IUPAC Name 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
Beilstein 3211892
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 272.3
Percent Purity 97%
Chemical Name or Material Anisoin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.