missing translation for 'onlineSavingsMsg'
Learn More

Aniracetam, Thermo Scientific™

 

Brand:  Thermo Scientific Alfa Aesar J61661.03

Quantity 1g

Product Code. 15404219

  • £84.30 / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • AMPA receptor potentiator
  • Slows the rate of ion-channel closing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

72432-10-1
MFCD00153767
14,662
Soluble in chloroform; Also soluble in ethanol; Insoluble in water
COC1=CC=C(C=C1)C(=O)N2CCCC2=O
219.24
CHEBI:47943
Aniracetam
C12H13NO3
1g
aniracetam, draganon, sarpul, 1-4-methoxybenzoyl-2-pyrrolidinone, ampamet, 1-4-methoxybenzoyl pyrrolidin-2-one, 1-p-anisoyl-2-pyrrolidinone, memodrin, aniracetamun inn-latin, aniracetam usan:inn:jan
ZXNRTKGTQJPIJK-UHFFFAOYSA-N
1-(4-methoxybenzoyl)pyrrolidin-2-one
2196
219.24
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.