missing translation for 'onlineSavingsMsg'
Learn More

Aniracetam, MedChemExpress

Product Code. 16417017
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16417017

Brand: Medchem Express HY10932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aniracetam(Ro 13-5057) is a nootropics and neuroprotective drug, which is selectively modulates the AMPA receptor and nAChR.

Aniracetam(Ro 13-5057) is a nootropics and neuroprotective drug, which is selectively modulates the AMPA receptor and nAChR.

Specifications

Chemical Name or Material Aniracetam
CAS 72432-10-1
Color White
Purity Grade Notes Research
Molecular Formula C12H13NO3
Quantity 5 g
Synonym Ro 13-5057
Solubility Information DMSO : ≥ 100 mg/mL (456.12 mM) ∣H2O : 0.33 mg/mL (1.51 mM; Need ultrasonic)
SMILES O=C(N1CCCC1=O)C2=CC=C(OC)C=C2
Molecular Weight (g/mol) 219.24
Formula Weight 219.24
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.