missing translation for 'onlineSavingsMsg'
Learn More

Aniracetam, MedChemExpress

Product Code. 16461129
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461129 1 g 1g
16417017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461129 Supplier Medchem Express Supplier No. HY10932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aniracetam(Ro 13-5057) is a nootropics and neuroprotective drug, which is selectively modulates the AMPA receptor and nAChR.

Aniracetam(Ro 13-5057) is a nootropics and neuroprotective drug, which is selectively modulates the AMPA receptor and nAChR.

Specifications

Chemical Name or Material Aniracetam
CAS 72432-10-1
Color White
Purity Grade Notes Research
Molecular Formula C12H13NO3
Quantity 1 g
Synonym Ro 13-5057
Solubility Information DMSO : ≥ 100 mg/mL (456.12 mM) ∣H2O : 0.33 mg/mL (1.51 mM; Need ultrasonic)
SMILES O=C(N1CCCC1=O)C2=CC=C(OC)C=C2
Molecular Weight (g/mol) 219.24
Formula Weight 219.24
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.