Learn More
STEMCELL Technologies Animal Component-Free Cell Dissociation Kit
Product Code. 15918682
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
15918682 1 Kit 1 set
1 options
Product Code. 15918682 Supplier STEMCELL Technologies Supplier No. 05426
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dissociation kit for human mesenchymal stem cells
  • Animal Component-Free Cell Dissociation Kit is optimized for dissociation and passaging of human stem and progenitor cells cultured in various media formulations
  • Animal Component-Free Cell Dissociation Kit includes ACF Enzymatic Dissociation Solution and ACF Enzyme Inhibition Solution
  • This kit was previously named MesenCult™-ACF Dissociation Kit
  • The name was changed to accommodate the use of this product with other cell types
Specifications
Cell Type Endothelial Cells, Endothelial Colony Forming Cells (ECFCs), HUVECs, Mesenchymal Cells, PSC-Derived, Mesenchymal Stem and Progenitor Cells, Myogenic Stem and Progenitor Cells
Quantity 1 Kit