Learn More
Invitrogen™ Anillin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595590
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell, rat PC-12 whole cell, mouse NIH/3T3 whole cell. ICC/IF: U2OS cell IP: U251 cell. Flow: U251 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes an actin-binding protein that plays a role in cell growth and migration, and in cytokinesis. The encoded protein is thought to regulate actin cytoskeletal dynamics in podocytes, components of the glomerulus. Mutations in this gene are associated with focal segmental glomerulosclerosis 8. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| Anillin | |
| Polyclonal | |
| Unconjugated | |
| ANLN | |
| 1110037A17Rik; 2900037I21Rik; actin-binding protein anillin; Anillin; anillin actin binding protein; anillin, actin binding protein; anillin, actin binding protein (scraps homolog, Drosophila); Anln; C78101; DKFZp779A055; FSGS8; RGD1561367; scra; Scraps | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 363031, 54443, 68743 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q8K298, Q9NQW6 | |
| ANLN | |
| A synthetic peptide corresponding to a sequence of human Anillin/ANLN (KEERDLWMQKLNQVLVDIRLWQPDACYK). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.