missing translation for 'onlineSavingsMsg'
Learn More

Aniline, TRC

Product Code. 30132670
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132670 1 g 1g
30133363 10 g 10g
18051320 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132670 Supplier Toronto Research Chemicals Supplier No. TRCA6624751G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6 H7 N
Molecular Weight (g/mol) 93.13
Synonym Aniline, Aminobenzene, Aminophen, Anyvim, Benzene, amino-, Blue Oil, C.I. 76000, NCI 176889, Phenylamine, Benzenamine
IUPAC Name aniline
SMILES Nc1ccccc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2
Formula Weight 93.0578
Percent Purity >95%
Chemical Name or Material Aniline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.