missing translation for 'onlineSavingsMsg'
Learn More

Aniline sulfate, 97%

Product Code. 11463017
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11463017

Brand: Thermo Scientific Alfa Aesar A15563.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 542-16-5 | (C6H5NH2)2·H2SO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-16-5
Molecular Formula (C6H5NH2)2·H2SO4
MDL Number MFCD00013110
Synonym Benzenamine sulfate 2:1

Specifications

Density 1.377
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 3729533
Solubility Information Soluble in water
Formula Weight 284.34
Percent Purity 98%
Chemical Name or Material Aniline sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.