missing translation for 'onlineSavingsMsg'
Learn More

Aniline hydrochloride, 99%

Product Code. p-60507 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10001760 100 g Plastic bottle 100g
10617132 500 g Plastic bottle 500g
13408388 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10001760 Supplier Thermo Scientific Acros Supplier No. 158411000
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-04-1 | C6H8ClN | 129.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-04-1
Molecular Formula C6H8ClN
Molecular Weight (g/mol) 129.59
MDL Number MFCD00012958
InChI Key MMCPOSDMTGQNKG-UHFFFAOYSA-N
Synonym aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline
PubChem CID 8870
IUPAC Name aniline;hydrochloride
SMILES [H+].[Cl-].NC1=CC=CC=C1

Specifications

Melting Point 196.0°C to 199.0°C
Color Cream-Yellow to White
Boiling Point 245.0°C
Flash Point 193°C
Loss on Drying 0.5 % max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5NH2·HCl
Quantity 100 g
Merck Index 15,652
Solubility Information Solubility in water: 1070g/L (25°C). Other solubilities: insoluble in ethyl ether and chloroform
Formula Weight 129.59
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Aniline hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.