missing translation for 'onlineSavingsMsg'
Learn More

Aniline, Extra Pure, SLR, Fisher Chemical

Product Code. 10660311 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10660311 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10660311 Supplier Fisher Chemical Supplier No. A/7240/PB17

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 62-53-3
Melting Point -6.2°C
Color Yellow
pH 8.8
Boiling Point 181°C
Packaging Plastic Coated Glass bottle
Molecular Formula C6H7N
MDL Number 7629
Quantity 2.5 L
Vapor Density 3.3
Formula Weight 93.13g/mol
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.