missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aniline, BAKER ANALYZED™ Reagent, J.T. Baker™
292.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Flash Point | 70.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 500 mL |
Specific Gravity | 1L = 1.02kg |
Formula Weight | 93.13 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Aniline |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P361-Take off immediately all contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'