missing translation for 'onlineSavingsMsg'
Learn More

Aniline, 99+%, ACS reagent

Product Code. 10216993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216993 2.5 L Glass bottle 2.5 L
10444321 5 mL Glass bottle 5mL
10137850 500 mL Glass bottle 500mL
10751832 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10216993 Supplier Thermo Scientific Acros Supplier No. 423420025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62-53-3 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6.2°C
Color Colorless to White
Density 1.0213g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 76°C
Ignition Residue 0.005 % max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NH2
Refractive Index 1.5845 to 1.5865
Quantity 2.5 L
Beilstein 12, 59
Fieser 06,21; 17,16
Merck Index 15, 652
Specific Gravity 1.0213
Solubility Information Solubility in water: 36g/L (20°C).
Viscosity 4.4 mPa.s (20°C)
Formula Weight 93.13
Vapor Pressure 0.5mmHg at 20°C
Percent Purity 99+%
Grade ACS Reagent
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Aniline
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.