missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anilazine-d4 (ring-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Anilazine-D4 |
| CAS | 1219802-20-6 |
| Molecular Formula | C9 D4 H Cl3 N4 |
| Quantity | 0.01 g |
| Synonym | 4,6-Dichloro-N-(2-chloro-3,4,5,6-tetradeuteriophenyl)-1,3,5-triazin-2-amine, Anilazine-d4 (ring-d4), Anilazine-d4, Anilazine-D4 (2-chlorophenyl-D4), Anilazine-D4, 4,6-Dichloro-N-(2-chlorophenyl-3,4,5,6-d4)-1,3,5-triazin-2-amine |
| InChI Formula | InChI=1 S/C9H5Cl3N4/c10-5-3-1-2-4-6(5)13-9-15-7(11)14-8(12)16-9/h1-4 H,(H,13,14,15,16)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Nc2nc(Cl)nc(Cl)n2)c(Cl)c1[2 H] |
| IUPAC Name | 4,6-dichloro-N-(2-chloro-3,4,5,6-tetradeuteriophenyl)-1,3,5-triazin-2-amine |
| Molecular Weight (g/mol) | 279.5465 |
| Formula Weight | 277.9831 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction