Learn More
Byotrol ANIGENE Professional Surface Disinfectant Cleaner - Defra-approved
Product Code. 31777207
SAFETY PRECAUTIONS DANGER: Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wash contaminated skin thoroughly after handling. IF ON SKIN: Wash with plenty of soap and water. Dispose of contents/container in accordance with national regulations.
Product Code. 31777207 Supplier Byotrol Supplier No. SKU056A
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SAFETY PRECAUTIONS DANGER: Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wash contaminated skin thoroughly after handling. IF ON SKIN: Wash with plenty of soap and water. Dispose of contents/container in accordance with national regulations.
Broad-spectrum disinfectant cleaner for animal health settings, helping control infections and protect staff and animals.
  • A trusted veterinary-grade surface disinfectant cleaner designed to support robust biosecurity in animal health environments from practices and referral hospitals to shelters and farms.
  • Approved for use against notifiable animal diseases giving animal health professionals confidence that their disinfection protocols meet recognized UK standards.
  • Effective against bacteria viruses fungi and spores helping to reduce the risk of cross-contamination and infection transmission between patients staff and environments.
  • Combines powerful cleaning with proven disinfection removing organic soiling while delivering reliable pathogen control to save time and simplify protocols.
  • Suitable for use across a wide range of hard surfaces including floors walls worktops kennels stables and transport areas commonly found in animal care settings.
  • Developed with animal health professionals in mind helping organizations demonstrate a clear commitment to infection prevention animal safety and staff wellbeing.
  • Part of the ANIGENE range trusted by vets animal charities and professionals worldwide for consistent performance technical support and sector-specific expertise.
  • Ideal for routine hygiene outbreak response and high-risk areas making it a dependable cornerstone of everyday cleaning and disinfection protocols.
Use biocides safely. Always read the product label and information before use. ANIGENE should be rinsed off food contact surfaces, food/water bowls and toys.