missing translation for 'onlineSavingsMsg'
Learn More

Anhydro Erythromycin A, TRC

Product Code. 17963215
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963215 1 mg 1mg
17902255 2.5 mg 2.5mg
17951025 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963215 Supplier Toronto Research Chemicals Supplier No. TRCA6389501MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23893-13-2
Molecular Formula C37 H65 N O12
Molecular Weight (g/mol) 715.91
Synonym Erythromycin, 9-deoxo-6,12-dideoxy-6,9:9,12-diepoxy-, 9-Deoxo-6,12-dideoxy-6,9:9,12-diepoxy-erythromycin, 6,15,16-Trioxatricyclo[10.2.1.11,4]hexadecane, erythromycin deriv., 6,9,12-Anhydroerythromycin A, 9-Deoxo-6,12-dideoxy-6,9:9,12-diepoxyerythromycin, Anhydroerythromycin, Anhydroerythromycin A, BRL 46355ER, EM 202, Erythromycin anhydride, Erythromycin Imp. D (EP), (1S,2R,3R,4S,5R,8R,9S,10S,11R,12R,14R)-9-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-5-ethyl-3hydroxy-2,4,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-6,15,16trioxatricyclo[10.2.1.11,4]hexadecan-7-one
IUPAC Name (1S,2R,3R,4S,5R,8R,9S,10S,11R,12R,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-5-ethyl-3-hydroxy-9-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-2,4,8,10,12,14-hexamethyl-6,15,16-trioxatricyclo[10.2.1.11,4]hexadecan-7-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)C[C@@H](C)[C@]5(O4)O[C@@]1(C)[C@H](O)[C@H]5C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C37H65NO12/c1-14-25-36(10)29(40)22(6)37(50-36)18(2)16-35(9,49-37)31(48-33-27(39)24(38(11)12)15-19(3)44-33)20(4)28(21(5)32(42)46-25)47-26-17-34(8,43-13)30(41)23(7)45-26/h18-31,33,39-41H,14-17H2,1-13H3/t18-,19-,20+,21-,22-,23+,24+,25-,26+,27-,28+,29-,30+,31-,33+,34-,35-,36-,37+/m1/s1
Formula Weight 715.4507
Percent Purity >95%
Chemical Name or Material Anhydroerythromycin A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.