missing translation for 'onlineSavingsMsg'
Learn More

Anhydro Chlortetracycline Hydrochloride, TRC

Product Code. 30137764
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137764 100 mg 100mg
30004740 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137764 Supplier Toronto Research Chemicals Supplier No. TRCA637740100MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65490-24-6
Molecular Formula C22 H21 Cl N2 O7 . Cl H
Molecular Weight (g/mol) 497.33
Synonym 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, hydrochloride (1:1), (4S,4aS,12aS)-, 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride, [4S-(4α,4aα,12aα)]- (9CI), 2-Naphthacenecarboxamide, 7-chloro-4-dimethylamino-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride (6CI), Anhydrochlortetracycline hydrochloride, (4S,4aS,12aS)-7-Chloro-4-(dimethylamino)-3,10,12,12a-tetrahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,11,12a-hexahydrotetracene-2-carboxamide hydrochloride
IUPAC Name (4S,4aS,12aS)-7-chloro-4-(dimethylamino)-3,10,12,12a-tetrahydroxy-6-methyl-1,11-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES Cl.CN(C)[C@H]1[C@@H]2CC3=C(C)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H21ClN2O7.ClH/c1-7-8-6-9-16(25(2)3)18(28)15(21(24)31)20(30)22(9,32)19(29)13(8)17(27)14-11(26)5-4-10(23)12(7)14;/h4-5,9,16,26,28-29,32H,6H2,1-3H3,(H2,24,31);1H/t9-,16-,22-;/m0./s1
Formula Weight 496.0804
Chemical Name or Material Anhydro Chlortetracycline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.