missing translation for 'onlineSavingsMsg'
Learn More

Anguizole, MedChemExpress

Product Code. 16491807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491807

Brand: Medchem Express HY13321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Anguizole is a small molecule inhibitor of HCV replication and alters NS4B(acute)s subcellular distribution.

Anguizole is a small molecule inhibitor of HCV replication and alters NS4B(acute)s subcellular distribution.

Specifications

Chemical Name or Material Anguizole
Physical Form Solid
CAS 442666-98-0
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H11ClF2N4O2S
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 50 mg/mL (122.31 mM; Need ultrasonic)
SMILES FC(Cl)(C1=CC(C2=CC=CO2)=NC3=CC(C(NCC4=CC=CS4)=O)=NN31)F
Molecular Weight (g/mol) 408.81
Formula Weight 408.81
Percent Purity 99.48%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.