missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL8 Mouse anti-Human, Clone: 2H9F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66641-1-IG-20UL
This item is not returnable.
View return policy
Description
The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.Specifications
| ANGPTL8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6UXH0 | |
| ANGPTL8 | |
| ANGPTL8 Fusion Protein Ag20708 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 2H9F9 | |
| Unconjugated | |
| ANGPTL8 | |
| Angiopoietin-like protein 8, ANGPTL8, Betatrophin, C19orf80, Lipasin, LOC55908, PRO1185, PVPA599, td26 | |
| Mouse | |
| Protein G | |
| RUO | |
| 55908 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction