missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122240
This item is not returnable.
View return policy
Description
ANGPTL6 Polyclonal Antibody for Western Blot, IHC (P)
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.
Specifications
| ANGPTL6 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL6 | |
| 6330404E11Rik; AGF; angiopoietin like 6; angiopoietin-like 6; angiopoietin-like protein 6; Angiopoietin-related growth factor; angiopoietin-related protein 5; Angiopoietin-related protein 6; Angptl6; ARP3; Arp5; UNQ152/PRO178 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 70726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8R0Z6 | |
| ANGPTL6 | |
| Recombinant protein Angiopoietin Like Protein 6. The antigen corresponds to amino acid range 238-457 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction