missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13986-1-AP-20UL
This item is not returnable.
View return policy
Tekniske data
| ANGPTL5 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL5 | |
| angiopoietin like 5, Angiopoietin like protein 5, Angiopoietin related protein 5, ANGPTL5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 253935 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86XS5 | |
| ANGPTL5 | |
| ANGPTL5 Fusion Protein Ag5053 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion