missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18374-1-AP-150UL
This item is not returnable.
View return policy
Description
Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.Specifications
| ANGPTL4 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL4 | |
| angiopoietin like 4, Angiopoietin like protein 4, Angiopoietin related protein 4, ANGPTL2, ANGPTL4, ARP4, FIAF, HFARP, NL2, PGAR, pp1158, PSEC0166, UNQ171/PRO197 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51129, 57875 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BY76, Q9Z1P8 | |
| ANGPTL4 | |
| ANGPTL4 Fusion Protein Ag13237 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction