missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529944
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Pig (84%), Sheep (84%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
Specifications
| ANGPTL4 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL4 | |
| 425O18-1; angiopoietin like 4; angiopoietin-like 4; angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; ANGPTL4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; FIAF; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; Pp1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 362850, 51129, 57875 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6TMA8, Q9BY76, Q9Z1P8 | |
| ANGPTL4 | |
| Recombinant protein encompassing a sequence within the center region of human Angiopoietin-like 4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction