missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578776
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat stomach tissue, mouse ovary tissue. IHC: mouse intestine tissue, rat intestine tissue, rat cardiac muscle tissue, human intetsinal cancer tissue.
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
Specifications
| ANGPTL2 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL2 | |
| AI593246; angiopoietin like 2; angiopoietin related protein 2; angiopoietin-like 2; angiopoietin-like protein 2; Angiopoietin-related protein 2; angiopoietin-related protein-2; Angptl2; ARP2; AW260363; HARP; MGC8889; UNQ170/PRO196 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171100, 23452, 26360 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q9R045, Q9UKU9 | |
| ANGPTL2 | |
| A synthetic peptide corresponding to a sequence in the middle region of human ANGPTL2 (275-312aa WRDCLQALEDGHDTSSIYLVKPENTNRLMQVWCDQRHD). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction