missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145335
This item is not returnable.
View return policy
Description
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
Specifications
| ANGPTL1 | |
| Polyclonal | |
| Unconjugated | |
| Angptl1 | |
| 2810039D03Rik; ANG3; ANG-3; angioarrestin; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); PSEC0154; UNQ162; UNQ162/PRO188 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100347829, 102184927, 679942, 72713, 9068 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| O95841, Q640P2 | |
| Angptl1 | |
| Recombinant Human ANG-3 protein, Phe271-Asp491 (Accession #O95841). | |
| 200 μL | |
| Primary | |
| Goat, Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction