missing translation for 'onlineSavingsMsg'
Learn More

Angiotensin I (human)

Product Code. 15454549
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454549 5 mg 5mg
15474549 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454549 Supplier Thermo Scientific Alfa Aesar Supplier No. J62102.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 484-42-4 | C62H89N17O14 | 1296.50 g/mol

Angiotensin I (human) stimulates the release of aldosterone, another hormone, from the adrenal cortex. It acts as a precursor to angiotensin II. It acts as endogenous peptide substrate for angiotensin converting enzyme (ACE), precursor to the vasoconstrictor peptide angiotensin II

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Angiotensin I (human) stimulates the release of aldosterone, another hormone, from the adrenal cortex. It acts as a precursor to angiotensin II. It acts as endogenous peptide substrate for angiotensin converting enzyme (ACE), precursor to the vasoconstrictor peptide angiotensin II

Solubility
Soluble to 1 mg/ml in water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-42-4
Molecular Formula C62H89N17O14
Molecular Weight (g/mol) 1296.50
MDL Number MFCD00133091
InChI Key ORWYRWWVDCYOMK-HBZPZAIKSA-N
Synonym angiotensin i, human angiotensin i, angiotensin i human, 5-ile-angiotensin i, angiotensin i rat, angiotensin i mouse, angiotensin 1 human, angiotensin i human, mouse, rat, pepsitensin, ile5-angiotensin i
PubChem CID 3081372
ChEBI CHEBI:2718
IUPAC Name (2S)-2-[(2S)-2-[(2S)-2-{[(2S)-1-[(2S)-2-[(2S,3S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-amino-3-carboxypropanamido]-5-[(diaminomethylidene)amino]pentanamido]-3-methylbutanamido]-3-(4-hydroxyphenyl)propanamido]-3-methylpentanamido]-3-(1H-imidazol-5-yl)propanoyl]pyrrolidin-2-yl]formamido}-3-phenylpropanamido]-3-(1H-imidazol-5-yl)propanamido]-4-methylpentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC1=CC=CC=C1)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N[C@@H](CC(C)C)C(O)=O

Specifications

Color White
Quantity 5 mg
Solubility Information Soluble to 1mg/ml in water.
Formula Weight 1296.5
Physical Form Powder
Chemical Name or Material Angiotensin I (human)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.