missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiotensin I Antibody (BGN/KA/4H) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£166.00 - £385.00
Specifications
| Antigen | Angiotensin I |
|---|---|
| Clone | BGN/KA/4H |
| Concentration | 1.0 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18053946
|
Novus Biologicals
NB100-62344-0.025MG |
0.025 mg |
£166.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18729403
|
Novus Biologicals
NB100-62344 |
0.1 mg | |||||||
Description
Angiotensin I Monoclonal specifically detects Angiotensin I in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
| Angiotensin I | |
| 1.0 mg/mL | |
| Unconjugated | |
| Mouse | |
| P01019 | |
| 183 | |
| Synthetic peptide corresponding to amino acids 40-49 of the angiotensinogen precursor (Swiss-Prot entry P01019) conjugated to HMW carrier protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BGN/KA/4H | |
| Monoclonal | |
| Purified | |
| RUO | |
| alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
| AGT | |
| IgG2a | |
| Protein A purified | |
| NB100-62344 is specific for Angiotensin I and does not react with angiotensin II, III or angiotensinogen in ELISA. Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. Angiotensin I is important in the Renin-Angiotensin-Aldosterone System (RAAS). RAAS is responsible for the regulation of blood pressure and water retention in the body. Angiotensin I is produced from a reaction between the renal enzyme Renin and angiotensinogen. It is converted by Angiotensin Converting Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title