missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like protein 8/Betatrophin Antibody (Regeneron patent anti-ANGPTL8), Janelia Fluor™ 549, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28032JF549
This item is not returnable.
View return policy
Description
Angiopoietin-like protein 8/Betatrophin Monoclonal antibody specifically detects Angiopoietin-like protein 8/Betatrophin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Angiopoietin-like protein 8/Betatrophin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599 | |
| ANGPTL8 | |
| 0.1 mL | |
| Cell Biology | |
| 55908 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Regeneron patent anti-ANGPTL8 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction