missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Angiopoietin-like protein 8/Betatrophin Antibody (Regeneron patent anti-ANGPTL8) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Angiopoietin-like protein 8/Betatrophin |
|---|---|
| Clone | Regeneron patent anti-ANGPTL8 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047412
|
Novus Biologicals™
NBP3-28032-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045945
|
Novus Biologicals™
NBP3-28032-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044392
|
Novus Biologicals™
NBP3-28032-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-like protein 8/Betatrophin Monoclonal antibody specifically detects Angiopoietin-like protein 8/Betatrophin in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Angiopoietin-like protein 8/Betatrophin | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cell Biology | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599 | |
| ANGPTL8 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Regeneron patent anti-ANGPTL8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q6UXH0 | |
| 55908 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title