missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 6/ANGPTL6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Angiopoietin-like Protein 6/ANGPTL6 Polyclonal specifically detects Angiopoietin-like Protein 6/ANGPTL6 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Angiopoietin-like Protein 6/ANGPTL6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | AGFAngiopoietin-related protein 5, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ARP5angiopoietin-related protein 6 |
| Host Species | Rabbit |
| Immunogen | The immunogen for Anti-Angiopoietin-like Protein 6/ANGPTL6 antibody is: synthetic peptide directed towards the C-terminal region of Human ANGL6 (NP_114123.2). Peptide sequence PESDHYRLRLGQYHGDAGDSLSWHNDKPFSTVDRDRDSYSGNCALYQRGG |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?