missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 4/ANGPTL4 Antibody (Nanyang Tech.U. patent anti-ANGPTL4) [Janelia Fluor™ 585], Novus Biologicals™
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | Angiopoietin-like Protein 4/ANGPTL4 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Nanyang Tech.U. patent anti-ANGPTL4 |
| Conjugate | Janelia Fluor 585 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein |
| Host Species | Human |
| Immunogen | ANGPTL4 |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?