missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 4/ANGPTL4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98267CL1
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 4/ANGPTL4 Polyclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| Produced in rabbits immunized with purified, recombinant Mouse Angiopoietin-like Protein 4/ANGPTL4 (Uniprot#: Q9Z1P8; Lys167-Ser410) | |
| 0.1 mL | |
| Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| 51129 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction