missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-like Protein 4/ANGPTL4 Antibody (547113) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30763011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763011 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763011 Supplier Novus Biologicals Supplier No. FAB97971MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiopoietin-like Protein 4/ANGPTL4
Applications ELISA
Classification Monoclonal
Clone 547113
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Host Species Rat
Immunogen CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein,, Gln24-Ser410, Accession # Q9Z1P8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51129
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.