missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 3/ANGPTL3 Mouse anti-Human, Mouse, Clone: 23D8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60244-50UL
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Monoclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Angiopoietin-like Protein 3/ANGPTL3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| ANG-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, FHBL2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 23D8 | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ANGPTL3 | |
| Recombinant human ANGPT5 (243-460aa) purified from E. coli. | |
| 50 μL | |
| Signal Transduction | |
| 27329 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction