missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 2/ANGPTL2 Antibody (RM0152-6F16), Alexa Fluor™ 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12040AF405
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 2/ANGPTL2 Monoclonal specifically detects Angiopoietin-like Protein 2/ANGPTL2 in Mouse samples. It is validated for Western Blot.
Specifications
| Angiopoietin-like Protein 2/ANGPTL2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANGPTL2 | |
| Mouse recombinant Angiopoietin-like Protein 2/ANGPTL2 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0152-6F16 | |
| Western Blot | |
| angiopoietin-like 2, Angiopoietin-like protein 2, angiopoietin-related protein 2, ARP2HARP, MGC8889 | |
| Rat | |
| Protein A or G purified | |
| Cardiovascular Biology | |
| 23452 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction