missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 1/ANGPTL1 Antibody (OTI1B2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71414
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 1/ANGPTL1 Monoclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Angiopoietin-like Protein 1/ANGPTL1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| ANG-3, ANG3angioarrestin, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, AngY, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162 | |
| Mouse | |
| 54.3 kDa | |
| 100 μg | |
| 9068 | |
| Human, Mouse | |
| Lyophilized |
| Western Blot, Immunohistochemistry | |
| OTI1B2 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ANGPTL1 | |
| Human recombinant protein fragment corresponding to amino acids 24-491 of human ANGPTL1(NP_004664) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction