missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-4 Antibody (RM0073-2H34), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21700F
This item is not returnable.
View return policy
Description
Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Angiopoietin-4 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ANGPT4 | |
| Mouse recombinant Angiopoietin-3 | |
| 0.1 mL | |
| 51378 | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0073-2H34 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Angiopoietin-3. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction