missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | Angiopoietin 2 |
|---|---|
| Concentration | 0.42 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Angiopoietin-2 (ANGPT2) is an antagonist of angiopoietin 1 (ANGPT1) and endothelial TEK tyrosine kinase (TIE-2, TEK). The protein disrupts the vascular remodeling ability of ANGPT1 and may induce endothelial cell apoptosis. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
| Angiopoietin 2 | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O15123, O35462, O35608 | |
| 11601, 285, 89805 | |
| Angiopoietin 2 Fusion Protein Ag17938 | |
| Primary | |
| -20°C | |
| ANGPT2 |
| 0.42 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AGPT2, ANG 2, ANG2, angiopoietin 2, ANGPT2 | |
| ANGPT2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title