missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-2 Antibody (Roche patent anti-ANG-2), PE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27955PE
This item is not returnable.
View return policy
Description
Angiopoietin-2 Monoclonal antibody specifically detects Angiopoietin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Angiopoietin-2 | |
| Monoclonal | |
| PE | |
| AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand | |
| ANGPT2 | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 285 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Roche patent anti-ANG-2 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction