Learn More
Invitrogen™ Angiopoietin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596841
Description
Immunogen sequence: REEEKPFRDC ADVYQAGFNK SGIYTIYINN MPEPKKVFCN MDVNGGGWTV IQHREDGSLD FQRGWKEYKM GFGNPSGEYW LGNEFIFAIT SQRQYMLRIE LMDWEGNRAY SQYDRFHIGN EKQNYRLYLK GHTGTAGKQS SLILHGADFS TKDADNDNCM CKCALMLTGG WWFDACGPSN LNGMFYTAGQ NHGKLNGIKW HYFKGPSYSL RSTTMMIRPL DF Positive Samples: A549, mouse lung.
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
Specifications
| Angiopoietin 1 | |
| Polyclonal | |
| Unconjugated | |
| Angpt1 | |
| 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11600, 284, 89807 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O08538, O35460, Q15389 | |
| Angpt1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.