Learn More
Angiopoietin 1 Mouse, anti-Human, Clone: 3C4A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68618-1-IG-150UL
Description
The Angiopoietin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Angiopoietin 1. This antibody recognizes human, mouse antigen. The Angiopoietin 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Angiopoietin 1 | |
| 3C4A3 | |
| AGP1, AGPT, ANG 1, ANG1, angiopoietin 1, ANGPT1, KIAA0003 | |
| 150 μL | |
| 284 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG1 |