missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (MM0019-26B10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-85464PCP
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal antibody specifically detects Angiopoietin-1 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Angiopoietin-1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Chicken | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0019-26B10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| Human recombinant human Angiopoietin-1 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 284 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction