missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Angiogenin Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5118949

 View more versions of this product

Product Code. 17720364

  • £430.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Angiogenin Polyclonal Antibody for Western Blot, IHC (P)

Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Angiogenin
Polyclonal
Unconjugated
ANG
AI385586; ALS9; ANG; Ang1; Angiogenin; angiogenin ribonuclease 1; angiogenin, ribonuclease A family, member 1; angiogenin, ribonuclease, RNase A family, 5; angiogenin-1; epididymis luminal protein 168; HEL168; MGC22466; MGC71966; RAA1; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5; RNASE4; RNASE5; Rnase5a
Rabbit
Antigen affinity chromatography, Protein A
RUO
305843
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
0
ANG
Recombinant protein Angiogenin.
200 μL
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.