missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGEL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA512329
This item is not returnable.
View return policy
Description
ANGEL2 Polyclonal Antibody for Western Blot, IHC (P)
ANGEL 2 is a member of the CCR4/nocturin family.
Specifications
| ANGEL2 | |
| Polyclonal | |
| Unconjugated | |
| ANGEL2 | |
| 2610307I21Rik; 5730410O10Rik; AI845692; angel homolog 2; angel homolog 2 (Drosophila); ANGEL2; Ccr4d; D1Ertd396e; D1Ertd654e; KIAA0759L; MNCb-4273; Protein angel homolog 2; RGD1305056 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 90806 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5VTE6 | |
| ANGEL2 | |
| KLH conjugated synthetic peptide between 108-138 amino acids from the central region of human ANGEL2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction