missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGEL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581567
This item is not returnable.
View return policy
Description
This antibody has specificity for Mouse ANGEL1.
ANGEL1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cis-Golgi network; cytosol; endoplasmic reticulum; nucleus; perinuclear region of cytoplasm; eukaryotic initiation factor 4E binding; protein domain specific binding.
Specifications
| ANGEL1 | |
| Polyclonal | |
| Unconjugated | |
| ANGEL1 | |
| 1110030H02Rik; angel homolog 1; angel homolog 1 (Drosophila); angel1; angel1 {ECO:0000312; Ccr4e; EMBL:AAI66827.1}; KIAA0759; LOW QUALITY PROTEIN: protein angel homolog 1; mKIAA0759; protein angel homolog 1; RGD1306238 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 68737 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q8VCU0 | |
| ANGEL1 | |
| A synthetic peptide corresponding to the center region of the Mouse ANGEL1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction