missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
80.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Used in feed additive, nutrient, firming agent and flavor enhancer. It is also used as a fermentation aid in the processing of beer and malt beverages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink |
Physical Form | Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 99.999% (Metal basis) |
Solubility Information | Soluble in water. |
IUPAC Name | manganese(2+) hydrate sulfate |
Formula Weight | 151.00 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Pink Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'