missing translation for 'onlineSavingsMsg'
Learn More

Anethole, MedChemExpress

Product Code. p-200098824
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16438496

Brand: Medchem Express HYB0900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Anethole is a type of aromatic compound that occurs widely in nature, widely used as a flavoring substance.

Anethole is a type of aromatic compound that occurs widely in nature, widely used as a flavoring substance.

Specifications

Chemical Name or Material Anethole
CAS 104-46-1
Color White
Purity Grade Notes Research
Molecular Formula C10H12O
Quantity 100 mg
Synonym Anise camphor p-Propenylanisole Isoestragole
Solubility Information DMSO : ≥ 100 mg/mL (674.76 mM)
SMILES C/C=C/C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 148.2
Formula Weight 148.2
Percent Purity 98.46%
Grade Research
Physical Form Solid-Liquid Mixture
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.