missing translation for 'onlineSavingsMsg'
Learn More

Anemarsaponin E, MedChemExpress

Product Code. 16461518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461518 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461518 Supplier Medchem Express Supplier No. HYN0813
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Anemarsaponin E is extracted from Anemarrhena asphodeloides Bunge and has anti-inflammatory activity.

Anemarsaponin E is extracted from Anemarrhena asphodeloides Bunge and has anti-inflammatory activity.

Specifications

Chemical Name or Material Anemarsaponin E
CAS 136565-73-6
Color White
Purity Grade Notes Research
Molecular Formula C46H78O19
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (106.94 mM; Need ultrasonic)
SMILES C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]3(OC)CC[C@H](C)CO[C@@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO)([H])C[C@@]1([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC2
Molecular Weight (g/mol) 935.1
Formula Weight 935.1
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.